Learn More
Description
Ideal for labeling of biological molecules (e.g. peptides, proteins, oligoes) for fluorescence imaging and other fluorescence-based biochemical analysis
- Chemical Formula: C40H47F3N2O12S2
- Mol. Weight: 868.93g/mol
- Purity: ≥90%
- Molar Extinction Co-efficient: ≥1,00,000 M-1cm-1
- Excitation (Max): 749nm
- Emission (Max): 776nm
- Solubility: DMSO, water
Application: Conjugation, In vivo imaging
Recommended Storage: Store in the dark at -20°C.
Specifications
Specifications
| Description | Cyanine 7 dyes are common near infrared red fluorophores used in in vivo imaging applications. |
| For Use With (Application) | Conjugation; in vivo Imaging |
| Quantity | 5 mg |
| Product Type | Fluorescent Dye 749-C7 bisAcid |
| Molecular Weight (g/mol) | 868.93g/mol |
| Purity | ≥90% |
| Content And Storage | Store in the dark at -20°C |
| Label or Dye | Fluorescent Dye 749-C7 bisAcid |
| Label Type | Other Labels or Dyes |
| Emission | 776 |
| Show More |
For research use only (RUO).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.