Learn More
Description
Compared with Alexa Fluor 750, Fluorescent Dye 750-I is much brighter with absorption at 633nm. Fluorescence emission of Fluorescent Dye 750-I Maleimide is well separated from that of other commonly used red fluorophores, such as TAMRA, Fluorescent Dye 647-I and R-phycoerythrin, making it ideal for multi-color labeling. Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis
- Spectral Equivalent: Alexa Fluor 750, Cy7, DyLight 755
- Purity: >90%
- Molar Extinction Co-efficient: >2,40,000 M-1cm-1
- Excitation (Max): 749nm
- Emission (Max): 775nm
- Solubility: DMSO, water
- Storage Buffer: DMSO
Application: Conjugation, Flow Cytometry
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Fluorescent dye that is water-soluble and forms stable thioether bond with free thiol groups. Compared with Alexa Fluor 750, Fluorescent Dye 750-I is much brighter with absorption at 633nm. Fluorescence emission of Fluorescent Dye 750-I Maleimide is well separated from that of other commonly used red fluorophores, such as TAMRA, Fluorescent Dye 647-I and R-phycoerythrin, making it ideal for multi-color labeling. |
| For Use With (Application) | Conjugation; Flow Cytometry |
| Quantity | 1 mg |
| Product Type | Fluorescent Dye 750-I Maleimide |
| Purity | >90% |
| Equivalence | Alexa Fluor 750, Cy7, DyLight 755 |
| Storage Buffer | DMSO |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. |
| Label or Dye | Fluorescent Dye 750-I Maleimide |
| Label Type | Other Labels or Dyes |
| Show More |
For research use only (RUO).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.