Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fluorescent Dye 750-I Succinimidyl Ester

Catalog No. 89139545 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-545 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-545 Supplier Abnova™ Supplier No. U0259
Add to Cart
Add to Cart

Water-soluble fluorescent dye forms stable peptide bond with protein amino groups

Compared with Alexa Fluor 750, Fluorescent Dye 750-I is much brighter with absorption at 633nm. Fluorescence emission of Fluorescent Dye 750-I Succinimidyl Ester is well separated from that of other commonly used red fluorophores, such as TAMRA, Fluorescent Dye 647-I and R-phycoerythrin, making it ideal for multi-color labeling. Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis

  • Spectral Equivalent: Alexa Fluor 750, Cy7, DyLight 755
  • Purity: >90%
  • Molar Extinction Co-efficient: >2,50,000 M-1cm-1
  • Excitation (Max): 749nm
  • Emission (Max): 775nm
  • Solubility: DMSO, water
  • Storage Buffer: DMSO

Application: Conjugation, Flow Cytometry

Recommended Storage: Store in desiccated environment at -20°C and avoid from light.

Specifications

Description Fluorescent dye that is water-soluble and forms stable peptide bond with protein amino groups. Compared with Alexa Fluor 750, Fluorescent Dye 750-I is much brighter with absorption at 633nm. Fluorescence emission of Fluorescent Dye 750-I Succinimidyl Ester is well separated from that of other commonly used red fluorophores, such as TAMRA, Fluorescent Dye 647-I and R-phycoerythrin, making it ideal for multi-color labeling.
For Use With (Application) Conjugation; Flow Cytometry
Quantity 1 mg
Product Type Fluorescent Dye 750-I Succinimidyl Ester
Purity >90%
Equivalence Alexa Fluor 750, Cy7, DyLight 755
Storage Buffer DMSO
Content And Storage Store the product in desiccated environment at -20°C and avoid from light.
Label or Dye Fluorescent Dye 750-I Succinimidyl Ester
Emission 775
Excitation 749
Show More Show Less

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.