Learn More
Description
The fluorescence emission maximum of Fluorescent Dye 790-I around 810nm is well separated from commonly used far-red fluorophores, including Fluorescent Dye 647-I, Fluorescent Dye 680-I or allophycocyanin (APC), facilitating the rapid growing NIR biological analysis. Fluorescent Dye 790-I is one of the brightest NIR fluorescent labeling dyes with fluorescence quantum yield close to 0.1. Its conjugates have been successfully used for NIR fluorescent probe-based in vivo imaging analysis. Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis
- Spectral Equivalent: Alexa Fluor 790, IRDye 800
- Molar Extinction Co-efficient: >2,50,000 M-1cm-1
- Excitation (Max): 783nm
- Emission (Max): 814nm
- Solubility: Water
- Storage Buffer: Water
Application: Conjugation, in vivo Imaging
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Water-soluble fluorescent dye. The fluorescence emission maximum of Fluorescent Dye 790-I around 810nm is well separated from commonly used far-red fluorophores, including Fluorescent Dye 647-I, Fluorescent Dye 680-I or allophycocyanin (APC), facilitating the rapid growing NIR biological analysis. Fluorescent Dye 790-I is one of the bightest NIR fluorescent labeling dyes with fluorescence quantum yield close to 0.1. Its conjugates have been successfully used for NIR fluorescent probe-based in vivo imaging analysis. |
| For Use With (Application) | Conjugation; in vivo Imaging |
| Quantity | 5 mg |
| Product Type | Fluorescent Dye 790-I Acid |
| Equivalence | Alexa Fluor 790, IRDye 800 |
| Storage Buffer | Water |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. |
| Label or Dye | Fluorescent Dye 790-I Acid |
| Label Type | Other Labels or Dyes |
| Emission | 814 |
| Show More |
For research use only (RUO).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.