Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fluorescent Dye 790-I Succinimidyl Ester

Catalog No. 89139552 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-552 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-552 Supplier Abnova™ Supplier No. U0266
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Water-soluble fluorescent dye forms stable peptide bond with protein amino groups

The fluorescence emission maximum of Fluorescent Dye 790-I around 810nm is well separated from commonly used far-red fluorophores, including Fluorescent Dye 647-I, Fluorescent Dye 680-I or allophycocyanin (APC), facilitating the rapid growing NIR biological analysis. Fluorescent Dye 790-I is one of the brightest NIR fluorescent labeling dyes with fluorescence quantum yield close to 0.1. Its conjugates have been successfully used for NIR fluorescent probe-based in vivo imaging analysis. Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis

  • Spectral Equivalent: Alexa Fluor 790, IRDye 800
  • Molar Extinction Co-efficient: >2,50,000 M-1cm-1
  • Excitation (Max): 783nm
  • Emission (Max): 814nm
  • Solubility: Water
  • Storage Buffer: Water

Application: Conjugation, in vivo Imaging

Recommended Storage: Store in desiccated environment at -20°C and avoid from light.

Specifications

Description Fluorescent dye that is water-soluble and forms stable peptide bond with protein amino groups. The fluorescence emission maximum of Fluorescent Dye 790-I around 810nm is well separated from commonly used far-red fluorophores, including Fluorescent Dye 647-I, Fluorescent Dye 680-I or allophycocyanin (APC), facilitating the rapid growing NIR biological analysis. Fluorescent Dye 790-I is one of the bightest NIR fluorescent labeling dyes with fluorescence quantum yield close to 0.1. Its conjugates have been successfully used for NIR fluorescent probe-based in vivo imaging analysis.
For Use With (Application) Conjugation; in vivo Imaging
Quantity 1 mg
Product Type Fluorescent Dye 790-I Succinimidyl Ester
Equivalence Alexa Fluor 790, IRDye 800
Storage Buffer Water
Content And Storage Store the product in desiccated environment at -20°C and avoid from light.
Label or Dye Fluorescent Dye 790-I Succinimidyl Ester
Emission 814
Excitation 783

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.