Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fluorescent Dye AMCA Phalloidin

Catalog No. 89139594 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
300 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-594 300 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-594 Supplier Abnova™ Supplier No. U0308
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorescence Dye AMCA with Phalloidin conjugate

Ideal for labeling via Phalloidin for fluorescence imaging and other fluorescence-based biochemical analysis.

  • Excitation (Max): 353nm
  • Emission (Max): 442nm
  • Solubility: DMSO
  • Storage Buffer: DMSO

Application: Conjugation

Recommended Storage: Store in desiccated environment at -20°C and avoid from light.

Specifications

Description Fluorescence Dye AMCA with Phalloidin conjugate.
For Use With (Application) Conjugation
Quantity 300 Reactions
Product Type Fluorescent Dye AMCA Phalloidin
Sufficient For 300 Reactions
Storage Buffer DMSO
Content And Storage Store the product in desiccated environment at -20°C and avoid from light.
Label or Dye Fluorescent Dye AMCA Phalloidin
Emission 442
Excitation 353

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.