Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fluorescent Dye Eu-Streptavidin

Catalog No. 89139596 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-596 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-596 Supplier Abnova™ Supplier No. U0310
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Fluorescent Dye Eu with Streptavidin conjugate

Fluorescent Dye Eu is optimized for TR-FRET applications with large Stokes shifts and extremely long emission half-lives when compared to traditional fluorophores such as Alexa Fluor™ or Cyanine dyes. Ideal for labeling via Streptavidin for fluorescence imaging and other fluorescence-based biochemical analysis. For most fluorescent streptavidin applications, a concentration of 1 to 20g/mL is sufficient. The optimal working dilution should be determined by the end user.

  • Typical Acceptor for TR-FRET: Alexa 647, APC, Cy5, DyLight 650, Fluorescence Dye 647-I, TF5
  • Molar Extinction Co-efficient: ≥22,000 M-1cm-1
  • Excitation (Max): 346nm
  • Emission (Max): 617nm
  • Solubility: Water
  • Storage Buffer: Water

Application: Conjugation, Time Resolved-Fluorescence Resonance Energy Transfer

Recommended Storage: Store in desiccated environment at -20°C and avoid from light.

Specifications

Description Fluorescent Dye Eu with Streptavidin conjugate. Fluorescent Dye Eu is optimized for TR-FRET applications with large Stokes shifts and extremely long emission half-lives when compared to traditional fluorophores such as Alexa Fluor™ or Cyanine dyes.
For Use With (Application) Conjugation; Time Resolved-Fluorescence Resonance Energy Transfer
Quantity 100 μg
Product Type Fluorescent Dye Eu-Streptavidin
Equivalence Alexa 647, APC, Cy5, DyLight 650, Fluorescence Dye 647-I, TF5
Storage Buffer Water
Content And Storage Store the product in desiccated environment at -20°C and avoid from light. Add ddH2O with 0.1% BSA into the vial to have 1 mg/mL stock solution. This reconstituted solution remains stable for 2 months when stored in the presence of 2 mM sodium azide and avoid from light at 4°C. For longer storage, divid the reconstituted solution into single-used aliquots, or add equal volume of glycerol without aliquoting, and avoid from light at -20°C.
Label or Dye Fluorescent Dye Eu-Streptavidin
Emission 617
Excitation 346

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.