Learn More
Description
Ideal for labeling via Phalloidin for fluorescence imaging and other fluorescence-based biochemical analysis.
- Excitation (Max): 546nm
- Emission (Max): 575nm
- Solubility: DMSO
- Storage Buffer: DMSO
Application: Conjugation
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Fluorescence Dye Tetramethylrhodamine with Phalloidin conjugate. |
| For Use With (Application) | Conjugation |
| Quantity | 300 Reactions |
| Product Type | Fluorescent Dye Tetramethylrhodamine Phalloidin |
| Sufficient For | 300 Reactions |
| Storage Buffer | DMSO |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. |
| Label or Dye | Fluorescent Dye Tetramethylrhodamine Phalloidin |
| Label Type | Classic Dyes |
| Emission | 575 |
| Show More |
For research use only (RUO).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.