Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ FN1 (Human) ELISA Kit

Catalog No. 89100746 Shop All Abnova Corporation Products
Catalog No. 89-100-746 Supplier Abnova™ Supplier No. KA3039
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • One 96-well plate pre-coated with anti-Human Fibronectin antibody, 96 (8 x 12) wells
  • Lyophilized Human Fibronectin standards, 10ng x 2
  • Sample / Standard diluent buffer, 30mL
  • Biotin conjugated anti-Human Fibronectin antibody (Concentrated), Dilution: 1:100, 130μL
  • Antibody diluent buffer, 12mL
  • Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100, 130μL
  • ABC diluent buffer, 12mL
  • TMB substrate, 10mL
  • Stop solution, 10mL
  • Wash buffer (25X), 30mL

Use to measure quantitatively human FN1.

This kit is based on sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 156 to 10000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description FN1 (Human) ELISA Kit is intended for the quantitative measurement of human FN1.
Detection Limit 10pg/mL
Detection Method Colorimetric
Includes One 96-well plate pre-coated with anti-Human Fibronectin antibody (96 (8 x 12) wells), Lyophilized Human Fibronectin standards (10ng x 2), Sample/Standard diluent buffer (30mL), Biotin conjugated anti-Human Fibronectin antibody (Concentrated), Dilution: 1:100 (130μL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100 (130μL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X): 30mL
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate
Sample Volume 100 μL
Sensitivity <10 pg/mL
Packaging 96-well
Content And Storage Store at 4°C for 4 months, or at -20°C for 8 months.
For Use With (Application) Quantification
Product Type FN1 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.