Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fn1 (Rat), Biotin, ELISA Kit

Catalog No. 89100311 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-311 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-311 Supplier Abnova™ Supplier No. KA0339
Add to Cart
Add to Cart

Includes

  • antibody coated microwells 96 wells
  • Lyophilized recombinant Rat Fn1 standard 10ng + 2 vials
  • Sample diluent buffer: 30mL
  • Biotinylated anti- Human HGF antibody 130μL
  • antibody diluent buffer 12mL
  • Avidin -Biotin -Peroxidase Complex (ABC) 130μL
  • ABC diluent buffer 12mL
  • TMB color developing agent 10mL
  • TMB stop solution 10mL

Sandwich Enzyme Immunoassay for the quantitative measurement of Human PGF

Specifications

Calibration Range 156 to 10000pg/mL
Conjugate Biotin
Cross Reactivity Rat
Description Fn1 (Rat) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of rat Fn1.
Detection Limit 15 pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Body fluid, cell culture supernatant, plasma, serum and tissue lysate
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type Fn1 (Rat) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.