Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ GBM Ab ELISA Kit

Catalog No. 89024558 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-558 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-558 Supplier Abnova™ Supplier No. KA1266

Includes

  • Micro titer strips Divisible microplate consisting of 12 modules of 8 wells each, coated with highly purified GBM. Ready to use. 12
  • Anti-GBM Calibrators (A-E) in a serum/buffer matrix (PBS, BSA, NaN3 <0.1% (w/w)) containing: 0; 20; 40; 80; 200U/mL Ready to use. 5 vials, 1.5mL each
  • Anti-GBM Controls in a serum/buffer matrix (PBS, BSA, NaN3 <0.1% (w/w)) positive (1) and negative (2), for the respective concentrations see the enclosed package insert. Ready to use. 2 vials, 1.5mL each
  • Sample buffer (Tris, NaN3 <0.1% (w/w)), yellow, concentrate (5x). 1 vial, 20mL
  • Enzyme Conjugate (PBS, Proclin 300 <0.5% (v/v)), (light red) containing polyclonal rabbit anti-human IgG; labelled with horseradish peroxidase. Ready to use. 1 vial, 15mL
  • TMB substrate solution TMB substrate solution. Ready to use. 1 vial, 15mL
  • Wash solution (PBS, NaN3 <0.1% (w/w)), concentrate (50x). 1 vial, 15mL

Indirect solid phase Enzyme Immunoassay (ELISA) for the quantitative measurement of IgG class autoantibodies to glomerular basement membrane (GBM) in Human Serum or Plasma

Specifications

Conjugate HRP
Cross Reactivity Human
Description Anti-GBM is an indirect solid phase enzyme immunoassay (ELISA) for the quantitative measurement of IgG class autoantibodies to glomerular basement membrane (GBM) in human serum or plasma.
Detection Method Colorimetric
Includes 1.5mL Anti-GBM calibrator, 1.5mL Anti-GBM controls, 20mL Sample Buffer, 15mL Enzyme Conjugate, 15mL TMB Substrate solution, 15mL stop solution, 12 micro titer strips
Quantity 1 Kit
Sample Type Plasma, Serum
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type GBM Ab ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.