Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ GC (Human) ELISA Kit

Catalog No. 89100242 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-242 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-242 Supplier Abnova™ Supplier No. KA0479
Add to Cart
Add to Cart

Includes

  • Human GC-Globulin Microplate: A 96-well polystyrene microplate coated with a polyclonal antibody against human GC-Globulin
  • Sealing Tapes: Precut, pressure-sensitive sealing tapes, which can be cut to fit the format of the individual assay
  • Human GC-Globulin Standard: Human GC-Globulin in a buffered protein base (Lyophilized), 150μg
  • Biotinylated Human GC-Globulin (Lyophilized)
  • EIA Diluent Concentrate (10x)
  • Wash Buffer Concentrate (20x)
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate
  • Chromogen Substrate
  • Stop Solution: A 0.5N hydrochloric acid

Use to measure quantitatively human GC. A competitive enzyme immunoassay.

This assay employs a quantitative competitive enzyme immunoassay technique that measures human GC-Globulin in less than 3 hours

Specifications

Calibration Range 0.098 to 100μg/mL
Conjugate Biotin
Cross Reactivity Human
Description GC (Human) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of human GC.
Detection Limit 0.09μg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Culture Supernatant, CSF, Plasma, Serum, Saliva, Milk
Sample Volume 25 μL
Packaging 96-well
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.