Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ GH (Human) ELISA Kit

Catalog No. 89100033 Shop All Abnova Corporation Products
Change view
Click to view available options
Calibration Range:
0 to 50ng/mL
0.1 to 12ng/mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Calibration Range
89-100-033 0 to 50ng/mL
89-100-206 0.1 to 12ng/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-033 Supplier Abnova™ Supplier No. KA0215
Add to Cart
Add to Cart

Includes

  • Sheep Anti-HGH-coated microtiter plate, 96 (8 x 12) wells
  • Reference standard set, containing 0, 2.5, 5, 10, 25, and 50ng/mL HGH, ready to use, 1 set
  • Enzyme Conjugate Reagent, 13mL
  • TMB Reagent (one step), 11mL
  • Stop Solution (1N HCl), 11mL

The GH (Human) ELISA Kit is based on the principle of a solid phase enzyme-linked immunosorbent assay (ELISA)

Specifications

Calibration Range 0 to 50ng/mL
Conjugate HRP
Cross Reactivity Human
Description GH (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of growth hormone.
Detection Limit 0.5ng/mL
Detection Method Colorimetric
Includes Antibody-Coated Wells (96 wells), Enzyme Conjugate Reagent (13mL), Reference Standard Set: 0, 2.5, 5, 10, 25, and 50 ng /mL, Lyophilized (1 set), TMB Reagent (11mL), Stop solution (11mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum
Sample Volume 50 μL
Sensitivity 0.5 ng/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.