Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ GH (Human) ELISA Kit

Catalog No. 89100206 Shop All Abnova Corporation Products
Change view
Click to view available options
Calibration Range:
0 to 50ng/mL
0.1 to 12ng/mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Calibration Range
89-100-206 0.1 to 12ng/mL
89-100-033 0 to 50ng/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-206 Supplier Abnova™ Supplier No. KA0521
Add to Cart
Add to Cart

Includes

  • Microtiter Plate, 96 wells
  • Conjugate, 12mL
  • hGH Standard, 12ng/mL, 1 vial
  • hGH Standard, 6ng/mL, 1 vial
  • hGH Standard, 2ng/mL, 1 vial
  • hGH Standard, 0.8ng/mL, 1 vial
  • hGH Standard, 0.2ng/mL, 1 vial
  • hGH Standard, 0ng/mL, 1 vial
  • Sample Diluent, 7mL
  • Substrate A, 10mL
  • Substrate B, 10mL
  • Stop Solution, 14mL
  • Wash Buffer (20X), 60mL

This hGH enzyme linked immunosorbent assay (ELISA) applies a technique called a quantitative sandwich immunoassay

Specifications

Calibration Range 0.1 to 12ng/mL
Conjugate HRP
Cross Reactivity Human
Description GH (Human) High sensitivity ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of GH.
Detection Limit 0.1ng/mL
Detection Method Colorimetric
Includes Antibody coated microwells (96 wells), hGH conjugate (15mL), hGH standards (12, 6, 2, 0.8, 0.2, 0ng/mL): Lyophilized, Sample diluent (7mL), Wash buffer 20X (60mL), Substrate A and B (10mL each), Stop solution (14mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum
Sample Volume 50 μL
Sensitivity 0.1 ng/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.