Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Giardia lamblia ELISA Kit

Catalog No. 89028824 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-824 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-824 Supplier Abnova™ Supplier No. KA2093

ELISA for the detection of Giardia lamblia cells in Human stool

Specifications

Conjugate HRP
Cross Reactivity Human
Description Giardia lamblia ELISA Kit is an enzyme linked immunosorbent assay for the detection of Giardia lamblia cells in human stool.
Detection Method Colorimetric
Includes 96-well break apart microtiter plate coated with Anti-G. lamblia Monoclonal Antibody, 100mL concentrated Wash Buffer (20X), 1 bottle, 60mL/1 bottle, 100mL stool Diluent (Blue), 1 bottle, 12mL/1 bottle 16mL HRP Conjugate (Green), 1 vial, 1.5mL/1 vial, 3.5mL positive control, 1 bottle, 14mL/1 bottle, 16mL TMB Substrate, 1 bottle, 15mL/1 bottle, 16mL stop solution, 1 unit plate cover
Quantity 96 Assays
Sample Type Stool
Sample Volume 400 μL
Content And Storage Store the kit at 4°C. Do not freeze.
For Use With (Application) Quantification
Product Type Giardia lamblia ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.