Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Global Histone H3-K27 Methylation Assay Kit

Catalog No. 89027143 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-143 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-143 Supplier Abnova™ Supplier No. KA0582
Add to Cart
Add to Cart

Use for measuring global Methylation of Histone H3-K27

Specifications

Description Global Histone H3-K27 Methylation Assay Kit is use for measuring global methylation of histone H3-K27.
Detection Method Colorimetric
Includes 10mL GG1 (10X Lysis Buffer), 16mL GG2 (Extraction Buffer), 28mL GG3 (Wash Buffer 10X), 1mL GG4 (Histone Buffer), 20mL GG5 (Blocking Buffer), 12mL GC6 (Antibody Buffer), 50μL GC7 (capture Antibody, 100μg/mL), 20
Quantity 1 Kit
Content And Storage Store GG8 and methylated H3-K27 control at -20°C away from light. Store GG5, GG7, GG9 and 8 well strips at 4°C away from light. Store all other components at room temperature.
For Use With (Application) Quantification
Product Type Global Histone H3-K27 Methylation assay kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.