Learn More
Specifications
Specifications
| Description | Global Histone H3-K27 Tri-Methylation Assay Kit is use for measuring global tri-methylation of histone H3-K27. |
| Detection Method | Colorimetric |
| Includes | 10mL GT1 (Wash Buffer 10X), 16mL GT2 (Extraction Buffer), 28mL GT3 (Wash Buffer 10X), 1mL GT4 (Histone Buffer), 20mL GT5 (Blocking Buffer), 12mL GT6 (Antibody Buffer), 50μL GT7 (capture Antibody, 100μg/mL), 20 |
| Quantity | 1 Kit |
| Content And Storage | Store GT8 and tri-methyl H3-K27 control at -20°C. Store GT5, GT7, GT9 and 8 well strips at 4°C away from light. Store all other components at room temperature. |
| For Use With (Application) | Quantification |
| Product Type | Global Histone H3-K27 Tri-Methylation assay kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.