Learn More
Specifications
Specifications
| Description | Global Histone H3-K4 Methylation Assay Kit is use for measuring global methylation of histone H3-K4. |
| Detection Method | Colorimetric |
| Includes | 10mL GD1 (Wash Buffer 10X), 16mL GD2 (Extraction Buffer), 28mL GD3 (Wash Buffer 10X), 1mL GD4 (Histone Buffer), 20mL GD5 (Blocking Buffer), 12mL GD6 (Antibody Buffer), 50μL GD7 (capture Antibody, 100μg/mL), 20 |
| Quantity | 1 Kit |
| Content And Storage | Store GD8 and methylated H3-K4 control at -20°C. Store GD5, GD7, GD9 and 8 well strips at 4°C away from light. Store other components at room temperature. |
| Form | Liquid |
| For Use With (Application) | Quantification |
| Product Type | Global Histone H3-K4 Methylation assay kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.