Learn More
Abnova™ Global Histone H3 Phosphorylation (Ser28) Assay Kit (Fluorometric)
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Cross Reactivity | Mammals |
| Description | Global Histone H3 Phosphorylation (Ser28) Assay Kit (Fluorometric) is use for measuring global phospho histone H3 (Ser28). |
| Detection Method | Fluorometric |
| Includes | 20mL F1 (Wash Buffer 10X), 12mL F2 (Antibody Buffer), 10μL F3 (detection Antibody, 1mg/mL), 24μL F4 (fluoro developer), 24μL F5 (fluoro enhancer), 8mL F6 (fluoro dilution), 20μL stand |
| Quantity | 1 Kit |
| Content And Storage | Store F3, F4 and standard control at -20°C. Store all other components at 4°C away from light. |
| For Use With (Application) | Quantification |
| Product Type | Global Histone H3 Phosphorylation (Ser28) assay kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.