Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Global Histone H4 Acetylation Assay Kit

Catalog No. 89027193 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-193 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-193 Supplier Abnova™ Supplier No. KA0634
Add to Cart
Add to Cart

Use for measuring global Acetylation of Histone H4

Specifications

Description Global Histone H4 Acetylation Assay Kit is use for measuring global acetylation of histone H4.
Detection Method Colorimetric
Includes 10mL GE1 (10X Lysis Buffer), 16mL GE2 (Extraction Buffer), 28mL GE3 (Wash Buffer 10X), 1mL GE4 (Histone Buffer), 20mL GE5 (Blocking Buffer), 12mL GE6 (Antibody Buffer), 50μL GE7 (capture Antibody, 100μg/mL), 20
Quantity 1 Kit
Content And Storage Store GE8 and acetylated histone H4 control at -20°C away from light. Store GE3, GE5, GE6, GE7, GE9 and 8 well strips at 4°C away from light. Store all other components at room temperature.
For Use With (Application) Quantification
Product Type Global Histone H4 Acetylation Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.