Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Global Pan-Methyl Histone H3-K36 Quantification Kit (Fluorometric)

Catalog No. 89027176 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-176 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-176 Supplier Abnova™ Supplier No. KA0615
Add to Cart
Add to Cart

Use for measuring global Mono-, Di- and Tri-Methylation of Histone H3-K36

Specifications

Description Global Pan-Methyl Histone H3-K36 Quantification Kit (Fluorometric) is use for measuring global mono-, di- and trimethylation of histone H3-K36.
Detection Method Fluorometric
Includes 20mL F1 (Wash Buffer 10X), 12mL F2 (Antibody Buffer), 10μL F3 (detection Antibody, 1mg/mL), 24μL F4 (fluoro developer), 24μL F5 (fluoro enhancer), 8mL F6 (fluoro-dilution), 20μL stand
Quantity 1 Kit
Content And Storage Store F3, F4 and standard control at -20°C. Store all other components at 4°C away from light.
For Use With (Application) Quantification
Product Type Global Pan Methyl Histone H3-K36 quantification kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.