Learn More
Description
- The Glutamate ELISA Kit is an enzyme immunoassay designed for the quantitative measurement of glutamate in urine and various biological samples.
- Assay Type: Quantitative
- Detection Method: Colorimetric
- Sensitivity: 0.17 &mu/mL
- Calibration/Standard Curve Range: 0.26 to 60 &mu/mL
- Sample Volume: 25 μL
- Label: Peroxidase-conjugated
- Precision: Intra-assay CV >6.8 % Inter-assay CV >16 %
- Spiking Recovery: 99%
- Applications: ELISA, Quantification
- Storage: Store at 4 °C
Specifications
Specifications
| Conjugate | HRP |
| Description | Glutamate ELISA Kit is an enzyme immunoassay for the quantitative measurement of glutamate in biological samples. |
| Detection Limit | 0.3μg/mL |
| Detection Method | Colorimetric |
| Includes | 1 x 96-well reaction plate, 1 x 4 adhesive foil, 1 x 20mL Wash Buffer concentrate, 1 x 12mL Enzyme Conjugate, 1 x 12mL Substrate, 1 x 12mL stop solution, 1 x 4mL standard A, 1 x 4mL standard B, 1 x 4mL standard C, 1 x 4mL standard D,1 x 4mL standard E, 1 |
| Quantity | 1 Kit |
| Sample Type | Plasma, Serum |
| Sample Volume | 50 μL |
| Content And Storage | Store the kit at 4°C |
| Form | Lyophilized |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.