Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ GSTM1 (Rat) ELISA Kit

Catalog No. 89100400 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-400 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-400 Supplier Abnova™ Supplier No. KA2135

Includes

  • Diluent Concentration (Running Buffer): 5X concentrated diluent running buffer
  • Wash Solution Concentrate: 20X concentrated wash solution
  • Enzyme-Antibody Conjugate 100X: Affinity purified anti-Rat GST Yb1 antibody conjugated with horseradish peroxidase in a stabilizing buffer
  • Chromogen-Substrate Solution
  • Stop Solution: 0.3M sulfuric acid. WARNING: Avoid contact with skin
  • Anti- Rat GST Yb1 ELISA micro plate: Twelve removable eight (8) well strips in well holder frame. Each well is coated with affinity purified anti-Rat GST Yb1
  • Rat GST Yb1 Calibrator: Rat GST Yb1 Calibrator

Measure rat Gstm1. Two-site enzyme linked immunoassay.

Specifications

Conjugate HRP
Cross Reactivity Rat
Description Gstm1 (Rat) ELISA Kit is a two-site enzyme linked immunoassay for the measurement of rat Gstm1.
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Biological Fluid
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store this kit at 4°C. Avoid repeated freezing and thawing.
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.