Learn More
Specifications
Specifications
| Description | HDAC Activity/Inhibition Assay Kit (Fluorometric) is use for measuring HDAC activity/inhibition. |
| Detection Method | Fluorometric |
| Includes | 28mL H1 (Wash Buffer 10X), 3mL H2 (HDAC Assay Buffer), 100μL H3 (Biotinylated HDAC Substrate), 100μL H4 (HDAC inhibitor, 0.5mM), 50μL H5 (HDAC assay standard 20μg/mL), 10μ |
| Quantity | 1 Kit |
| Content And Storage | Store H3, H4, H5 and H7 at -20°C away from light. Store H1, H6, Fluoro-Developer and 8-Well Strips at 4°C away from light. Store H2 at room temperature. |
| For Use With (Application) | Functional Study |
| Product Type | HDAC activity/inhibition assay kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.