Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Helicobacter pylori IgM ELISA Kit

Catalog No. 89100028 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-028 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-028 Supplier Abnova™ Supplier No. KA0221
Add to Cart
Add to Cart

Includes

  • Purified H. pylori antigen coated microtiter plate, 96 wells, 1 plate
  • Enzyme Conjugate Reagent (red color), 13mL
  • Sample Diluent (blue color), 22mL
  • Negative Control, 100μL
  • Cut-off Calibrator, H. pylori IgM EIA Index = 1, 100μL
  • Positive Control, 100μL
  • Wash Buffer (20x), 50mL
  • TMB Reagent (One-Step), 11mL
  • Stop Solution(1N HCl), 11mL

Use to detect Helicobacter pylori IgM. Sandwich enzyme immunoassay.

The Helicobacter pylori IgM ELISA Kit is intended for use in evaluating the serologic status to H. pylori infection in human with gastrointestinal symptoms

Specifications

Conjugate HRP
Cross Reactivity Helicobacter pylori
Description Helicobacter pylori IgM ELISA Kit is an enzyme immunoassay for the detection of Helicobacter pylori IgM .
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Qualitative
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.