Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Hemoglobin (Human) ELISA Kit

Catalog No. 89100352 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-352 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-352 Supplier Abnova™ Supplier No. KA2131
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Diluent Concentrate (Running buffer): One bottle containing 5X concentrated diluent running buffer
  • Wash Solution Concentrate, 20X
  • Enzyme-Antibody Conjugate 100X: One vial containing affinity purified anti-Human Hemoglobin antibody conjugated with horseradish peroxidase in a stabilizing buffer
  • Chromogen-Substrate Solution
  • Stop Solution: One vial containing 0.3M sulfuric acid. WARNING: Avoid contact with skin
  • Anti-Human Hemoglobin ELISA Micro Plate: Twelve removable eight (8) well strips in well holder frame. Each well is coated with affinity purified anti-Human Hemoglobin
  • Human Hemoglobin Calibrator

Measure human Hemoglobin. Two-site enzyme linked immunoassay.

In this assay the Hemoglobin present in samples reacts with the anti-Hemoglobin antibodies which have been adsorbed to the surface of polystyrene microtitre wells

Specifications

Calibration Range 6.25 to 200ng/mL
Conjugate HRP
Cross Reactivity Human
Description Hemoglobin (Human) ELISA Kit is a two-site enzyme linked immunoassay for the measurement of human Hemoglobin.
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store the calibrator at 4°C.
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.