Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Hepatitis A Virus Ab ELISA Kit

Catalog No. 89100160 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-160 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-160 Supplier Abnova™ Supplier No. KA0284
Add to Cart
Add to Cart

Detect antibodies to hepatitis A virus. Sandwich enzyme immunoassay.

Specifications

Conjugate HRP
Description Hepatitis A virus antibody ELISA Kit is a sandwich enzyme immunoassay for the detection of antibody to hepatitis A virus.
Detection Method Colorimetric
Quantity 1 Kit
Sample Type Plasma, Serum
Sample Volume 10 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Qualitative
Product Type Hepatitis A virus Ab ELISA Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.