Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Hepatitis B surface antigen Ab ELISA Kit

Catalog No. p-4396512 Shop All Abnova Corporation Products
Change view
Click to view available options
Product Type:
Hepatitis B Surface Antigen Ab ELISA Kit
Hepatitis B Surface Antigen ELISA Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
89-022-765 Hepatitis B Surface Antigen Ab ELISA Kit
89-100-158 Hepatitis B Surface Antigen ELISA Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-022-765 Supplier Abnova™ Supplier No. KA0287
Only null left
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the measurement of antibodies to Hepatitis B surface antigen

Specifications

Calibration Range 0 to 1000mIU/mL
Conjugate HRP
Description Hepatitis B surface antigen antibody ELISA Kit is a sandwich enzyme immunoassay for the measurement of antibodies to hepatitis B surface antigen.
Detection Limit 10mIU/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 6mL HBsAg -Peroxidase solution, 1mL Anti-HBs positive control, 1.5mL HB negative control, 10mL TMB Substrate solution A, 10mL TMB Substrate solution B, 50mL Conc. washing solution D (20X), 10mL 2N Sulfuric acid
Quantity 1 Kit
Sample Type Plasma, Serum
Sample Volume 50 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Qualitative
Product Type Hepatitis B Surface Antigen Ab ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.