Learn More
Specifications
Specifications
| Conjugate | HRP |
| Description | Hepatitis B virus core antigen antibody ELISA Kit is a sandwich enzyme immunoassay for the measurement of antibodies to hepatitis B virus core antigen. |
| Detection Method | Colorimetric |
| Quantity | 1 Kit |
| Sample Type | Plasma, Serum |
| Sample Volume | 50 μL |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Qualitative |
| Product Type | Hepatitis B virus core antigen Ab ELISA Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.