Learn More
Specifications
Specifications
| Conjugate | HRP |
| Cross Reactivity | Viruses |
| Description | Hepatitis B virus IgM ELISA Kit is a sandwich enzyme immunoassay for the measurement of IgM antibodies to hepatitis B virus. |
| Detection Method | Colorimetric |
| Quantity | 1 Kit |
| Sample Type | Plasma, Serum |
| Sample Volume | 100 μL |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Qualitative |
| Product Type | Hepatitis B virus IgM ELISA Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.