Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Hepatitis C Virus Ab ELISA Kit

Catalog No. 89100716 Shop All Abnova Corporation Products
Change view
Click to view available options
No. of Assays:
96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Assays
89-100-716 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-716 Supplier Abnova™ Supplier No. KA2534
Add to Cart
Add to Cart

Includes

  • HCV Antigens Plate, 1 plate
  • Conc. HCV Ag
  • HRPO Conjugate, 1.8mL
  • Anti-HCV Positive Control, 2mL
  • HCV Negative Control, 3mL
  • Conjugate Diluent, 24mL
  • TMB Substrate Solution A, 12mL
  • TMB Substrate Solution B, 12mL
  • Conc. Washing Solution D (20X), 110mL
  • 1N H2SO4, 12mL

In-vitro qualitative detection of Antibody to Hepatitis C virus (anti-HCV) in human serum or plasma.

This kit adopts the “direct sandwich principle” as the basis for the assay to detect antibodies to Hepatitis C virus (anti-HCV) in human serum or plasma

Specifications

Conjugate HRP
Description Hepatitis C virus antibody ELISA Kit is for in-vitro qualitative detection of Antibody to Hepatitis C virus (anti-HCV) in human serum or plasma.
Detection Method Colorimetric
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum
Sample Volume 100 μL
Sensitivity 100%
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Hepatitis C virus Ab ELISA Kit
Specificity 100%
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.