Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Herpes Simplex Virus I IgG (Mouse/Rat) ELISA Kit

Catalog No. 89028147 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-147 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-147 Supplier Abnova™ Supplier No. KA1046
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indirect Enzyme Immunoassay for the measurement of Mouse/Rat HSV-1 specific IgG class antibodies in Serum and Plasma

Specifications

Conjugate HRP
Cross Reactivity Mouse, Rat
Description Herpes Simplex Virus I IgG (Mouse/Rat) ELISA Kit is an indirect enzyme immunoassay for the measurement of mouse/rat HSV-1 specific IgG class antibodies in serum and plasma.
Detection Limit 90IU/mL
Detection Method Colorimetric
Includes 12 x 8 x 1 Microwell, 22mL sample diluent, 1.5mL calibrator, 1.5mL positive control, 1.5mL negative control, 12mL Enzyme Conjugate, 12mL TMB Chromogenic Substrate, 12mL stop solution, 25mL washing concentrate 20x
Quantity 1 Kit
Sample Type Serum
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Qualitative
Product Type Herpes Simplex Virus I IgG (Mouse/Rat) ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.