Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Herpes Simplex Virus I IgM ELISA Kit

Catalog No. 89100020 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-020 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-020 Supplier Abnova™ Supplier No. KA0230
Add to Cart
Add to Cart

Includes

  • Microtiter Wells: purified HSV-1 antigen coated wells (12 x 8 wells)
  • Enzyme Conjugate Reagent (red color): 1 vial (12mL)
  • Sample Diluent (blue color): 1 bottle (22mL)
  • Negative Control: Range stated on label. Natural cap (150μL/vial)
  • Cut-off Calibrator: Yellow cap. HSV-1 IgM Index = 1 (150μL/vial)
  • Positive Control: Range stated on label. Red cap. (150μL/vial)
  • Wash Buffer Concentrate (20x): 1 bottle (50mL)
  • TMB Reagent (One-Step): 1 vial (11mL)
  • Stop Solution (1N HCl): Natural cap. 1 vial (11mL)

Use to detect Herpes Simplex Virus I IgM. Sandwich enzyme immunoassay.

Specifications

Conjugate HRP
Cross Reactivity Viruses
Description Herpes Simplex Virus I IgM ELISA Kit is an enzyme immunoassay for the detection of Herpes Simplex Virus I IgM.
Detection Limit 93.00%
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum
Sample Volume 100 μL
Sensitivity 93.90%
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Qualitative
Product Type ELISA Kit
Specificity 94.90%
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.