Learn More
Specifications
Specifications
| Calibration Range | 10 to 1000AU |
| Conjugate | HRP |
| Detection Limit | 2AU |
| Detection Method | Colorimetric |
| Includes | 96-well ELISA break away strip coated with specific Antigen, 1.3mL/ each vial standard 1to 6, 0.2mL Anti-Human IgG Antibodies labelled with HRP, 125mL Wash Buffer concentrate, 125mL Dilution Buffer, 15mL chromogenic Substrate, 30mL stop solution |
| Quantity | 1 Kit |
| Sample Type | Cerebrospinal Fluid, Serum |
| Sample Volume | 100 μL |
| Content And Storage | 4°C |
| For Use With (Application) | For the quantitative measurement of IgG class autoantibodies against Herpes Simplex Virus I/II |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.