Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ HGF (Human), Biotin, ELISA Kit

Catalog No. 89100309 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-309 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-309 Supplier Abnova™ Supplier No. KA0343
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • antibody coated microwells 96 wells
  • Lyophilized recombinant Human HGF standard 10ng + 2 vials
  • Sample diluent buffer: 30mL
  • Biotinylated anti- Human HGF antibody 130μL
  • antibody diluent buffer 12mL
  • Avidin -Biotin -Peroxidase Complex (ABC) 130μL
  • ABC diluent buffer 12mL
  • TMB color developing agent 10mL
  • TMB stop solution 10mL

Sandwich Enzyme Immunoassay for the quantitative measurement of Human PGF

Specifications

Calibration Range 31.2 to 2000 pg/mL
Conjugate Biotin
Cross Reactivity Human
Description HGF (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human HGF.
Detection Limit 5 pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell culture supernatant, plasma (EDTA, Citrate) and serum
Sample Volume 50 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type HGF (Human) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.