Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ HHV-6 IgG ELISA Kit

Catalog No. 89028297 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-297 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-297 Supplier Abnova™ Supplier No. KA1457
Add to Cart
Add to Cart

Includes

  • ELISA strips coated with native antigen, 96-wells
  • Standard 1, Ready to use, 1.3mL
  • Standard 2, Ready to use, 1.3mL
  • Negative control serum, Ready to use, 1.3mL
  • Anti-human IgG antibodies labeled with horseraddish peroxidase (Px-conjugate), Ready to use, 1.3mL
  • Wash buffer 10x concentrated, 125mL
  • Dilution buffer, Ready to use, 100mL
  • Chromogenic substrate (TMB substrate), Ready to use, 13mL
  • Stop solution, Ready to use, 15mL
  • Sealable pouch for unused strips, 1 bag

Indirect Solid Phase ELISA for the semi-quantitative measurement of IgG class autoantibodies against Human Herpesvirus 6

Specifications

Conjugate HRP
Description Human Herpesvirus 6 IgG ELISA Kit is a solid phase enzyme immunoassay for the semiquantitative measurement of IgG class autoantibodies against Human Herpesvirus 6.
Detection Method Colorimetric
Includes 96-well ELISA strip (clear) coated with native Antigen, 1.2mL calibrator, 1.2mL positive control Serum, 1.2mL negative control Serum, 0.2mL Anti-Human IgG Antibodies labeled with HRP 50X concentrated (Px Conjugate), 125mL Wash Buffer 10X concentrate, 125mL Dilution Buffer (DB), 15mL Chromogenic Substrate (TMB Substrate), 30mL stop solution
Quantity 1 Kit
Sample Type Serum
Content And Storage Store at 4°C on dry atmosphere away from light.
For Use With (Application) Quantification
Product Type HHV-6 IgG ELISA kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.