Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ HIF1A Transcription Factor Assay Kit

Catalog No. 89024607 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-607 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-607 Supplier Abnova™ Supplier No. KA1321
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HIF1A Transcription Factor Assay Kit is a colorimetric method for determining HIF-1-alpha DNA binding activity.

  • A non-radioactive, sensitive method for detecting specific transcription factor DNA binding activity in nuclear extracts and whole cell lysate
  • 96-well enzyme-linked immunosorbent assay (ELISA) replaces the cumbersome radioactive electrophoretic mobility shift assay (EMSA)
  • Kit detects human, murine, and rat HIF-1a
  • Store kit at -20°C prior to use

Specifications

Description HIF1A Transcription Factor Assay Kit is a colorimetric method for determining HIF-1-alpha DNA binding activity.
Immunoassay Kit Format ELISA (Standard)
Product Type HIF1A Transcription Factor Assay Kit
Sufficient For 15 Reactions
For Use With (Application) Functional Study
Quantity 1 Kit
Storage Requirements Store the kit at -20°C temporarily. For long term storage, the positive control should be stored at -80°C. For best results, store the components as described in the protocol.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.