Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Histone Demethylase (H3-K4 Specific) Activity/Inhibition Assay Kit

Catalog No. 89027137 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-137 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-137 Supplier Abnova™ Supplier No. KA0573
Add to Cart
Add to Cart

Used for measuring activity/inhibition of H3-K4 specific Histone Demethylases using cell/ tissue extracts

Specifications

Description Histone Demethylase (H3-K4 Specific) Activity/Inhibition Assay Kit is use for measuring activity/inhibition of H3-K4 specific histone demethylases using cell/ tissue extracts.
Includes 22mL GM1 (Wash Buffer 10X), 3mL GM2 (DNA binding solution), 16μL GM3 (Methylated DNA control, 100μg/mL), 20mL GM4 (block solution), 10mL GM7 (developing solution), 6mL GM8 (stop solution), 12 8-well sample strip (with f
Quantity 1 Kit
Content And Storage Store HD3, HD4, HD6 and HD7 at -20°C away from light. Store all other components at 4°C away from light.
For Use With (Application) Functional Study
Product Type Histone Demethylase (H3-K4 Specific) Activity/Inhibition Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.