Learn More
Specifications
Specifications
| Description | Histone Demethylase LSD1 Activity/Inhibition Assay Kit is use for measuring cellular LSD1 activity/inhibition. |
| Includes | 30mL HG1 (Wash Buffer 10X), 4mL HG2 (LSD1 Assay Buffer), 100μL HG3 (LSD1 Substrate), 50μL HG4 (LSD1 standard, 20μg/mL), 10μL HG5 (capture Antibody 1000μg/mL), 16 |
| Quantity | 1 Kit |
| Content And Storage | Store HG3, HG4, and HG6 -20°C away from light. Store all other components at 4°C away from light. |
| For Use With (Application) | Functional Study |
| Product Type | Histone Demethylase LSD1 activity/inhibition assay kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.