Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Histone H3 (Total) Quantification Kit (Fluorometric)

Catalog No. 89027186 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-186 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-186 Supplier Abnova™ Supplier No. KA0625
Add to Cart
Add to Cart

Includes

  • F1 (10X wash buffer) 20mL
  • F2 (antibody buffer) 12mL
  • F3 (detection antibody, 1mg/ml) 10μL
  • F4 (fluoro developer) 24μL
  • F5 (fluoro enhancer) 24μL
  • F6 (fluoro dilution) 8mL
  • Standard control (100μg/ml) 20μL
  • 8 well sample strips (with frame) 9
  • 8 well standard control strips 3

Histone H3 (Total) Quantification Kit (Fluorometric) is designed for quantifying levels of histone H3 proteins independent of its modified state and can also be used for normalizing the modified histone H3 content of samples when run in parallel with other histone modification quantification kit series.

  • Quick and efficient procedure which can be finished within 2.5 hours
  • Innovative colorimetric assay without the need for radioactivity, electrophoresis, or chromatography
  • Specifically captures histone H3 with the detection limit as low as 2 ng/well and detection range from 10 ng-1 μg/well of histone extracts
  • The unmodified histone H3 control is conveniently included for quantification of the amount of total histone H3
  • Strip microplate format makes the assay flexible, manual or high throughput
  • Simple, reliable, and consistent assay conditions
  • Kit is shipped in two parts, one at ambient room temperature and the second on frozen ice packs at 4°C
  • Kit configured for research use only and is not intended for diagnostic and therapeutic application

Specifications

Description Histone H3 (Total) Quantification Kit (Fluorometric) is designed for quantifying levels of histone H3 proteins independent of its modified state and can also be used for normalizing the modified histone H3 content of samples when run in parallel with other histone modification quantification kit series.
Detection Method Fluorometric
Product Type Histo ne H3 (to tal) quantification Kit
For Use With (Application) Quantification
Quantity 96 Assays
Storage Requirements Store F3, F4 and Standard control at -20°C. Store all other components at 4°C away from light.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.