Learn More
Abnova™ Histone Methyltransferase Activity/Inhibition Assay Kit (H3-K27)
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Description | Histone Methyltransferase Activity/Inhibition Assay Kit (H3-K27) is use for measuring HMT activity/inhibition. |
| Includes | 22mL HE1 (Wash Buffer 10X), 3mL HE2 (Histone Assay Buffer), 50μL HE3 (Adomet), 200μL HE4 (Biotinylated Substrate, 25μg/mL), 20μL HE5 (HMT standard, 10μg/mL), 50 |
| Quantity | 1 Kit |
| Content And Storage | Store HE3, HE4, HE5, HE7, and control enzyme at -20°C away from light. Store all other components at 4°C away from light. |
| For Use With (Application) | Functional Study |
| Product Type | Histone Methyltransferase activity/inhibition assay kit (H3-K27) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.