Learn More
Abnova™ Histone Methyltransferase Activity/Inhibition Assay Kit (H3-K4)
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Description | Histone Methyltransferase Activity/Inhibition Assay Kit (H3-K4) is use for measuring HMT activity/inhibition. |
| Includes | 22mL HM1 (Wash Buffer 10X), 3mL HM2 (Histone Assay Buffer), 50μL HM3 (Adomet), 200μL HM4 (Biotinylated Substrate, 25μg/mL), 20μL HM5 (HMT standard, 10μg/mL), 50 |
| Quantity | 1 Kit |
| Content And Storage | Store HM3, HM4 HM5, HM7 and control enzyme at -20°C away from light. Store all other components at 4°C away from light. |
| For Use With (Application) | Functional Study |
| Product Type | Histone Methyltransferase activity/inhibition assay kit (H3-K4) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.