Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Hmox1 (Rat) ELISA Kit

Catalog No. 89028171 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-171 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-171 Supplier Abnova™ Supplier No. KA1019
Add to Cart
Add to Cart

Colorimetric immunometric Enzyme Immunoassay kit for the quantitative measurement of Rat Hmox1 in cell lysates, Plasma, Serum and tissue

Specifications

Calibration Range 0.195 to 12.5ng/mL
Conjugate HRP
Cross Reactivity Rat
Description Hmox1 (Rat) ELISA Kit is a colorimetric immunometric enzyme immunoassay kit for the quatitative measurement of rat Hmox1 in cell lysates, plasma, serum, and tissue.
Detection Limit 0.036ng/mL
Detection Method Colorimetric
Includes 96-well microwell, 10mL 5X HO-1 extraction reagent, 25μL recombinant Rat HO-1 standard, 50mL sample diluent, 100mL Wash Buffer, 10mL Anti-HO-1 Antibody, 10mL Anti-HO-1 Diluent, 10mL Anti-Rabbit IgG, HRP Conjugate, 10mL Anti-Rabbit IgG, H
Quantity 1 Kit
Sample Type Cell Lysate, Plasma, Serum, Tissue
Sample Volume 100 μL
Content And Storage Store Standard at -20°C. Store all other components at 4°C.
For Use With (Application) Quantification
Product Type HMOX1 (Rat) ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.