Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ HP (Bovine) ELISA Kit

Catalog No. 89100617 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Urine
Plasma, Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-617 Plasma, Serum
89-028-601 CCS, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-617 Supplier Abnova™ Supplier No. KA2421
Only null left
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Bovine HP

Specifications

Conjugate HRP
Cross Reactivity Bovine
Description HP (Bovine) ELISA Kit is for the quantitative measurement of HP in plasma, serum and milk.
Detection Limit 7.8 to 250ng/mL
Detection Method Colorimetric
Includes Anti-bovine haptoglobin antibody coated microtiter plate 96 wells, Enzyme Conjugate Reagent (11mL), Reference standard (lyophilized), 2.5μg/mL (1 vial ), Wash Buffer (10x stock) (60mL), 10x Diluent (25mL), TMB Reagent (One-Step) (11mL), Stop Solution (1N HCl) (11mL)
Quantity 96 Assays
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type HP (Bovine) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.