Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ HP (Dog) ELISA Kit

Catalog No. 89100628 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Plasma, Serum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-628 Plasma, Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-628 Supplier Abnova™ Supplier No. KA2432
Add to Cart
Add to Cart

Includes

  • Anti-dog haptoglobin antibody coated plate, 96 wells (12 detachable strips of 8)
  • cTnI Stock: Lyophilized bovine cTnI, Reconstitute with 0.40mL H2O
  • Enzyme Conjugate Reagent, 11mL
  • Reference standard (lyophilized), containing 2μg/mL dog haptoglobin
  • 20x Wash Solution, 50mL
  • TMB Reagent, 11mL
  • Stop Solution (1N HCl), 11mL

Use to measure quantitatively HP in plasma snd serum.

The dog haptoglobin ELISA is based on a solid phase enzymelinked immunosorbent assay (ELISA)

Specifications

Calibration Range 1.95 to 125ng/mL
Conjugate HRP
Cross Reactivity Dog
Description HP (Dog) ELISA Kit is for the quantitative measurement of HP in plasma snd serum.
Detection Limit 1.95 to 125ng/mL
Detection Method Colorimetric
Includes Anti-dog haptoglobin antibody coated plate (96 wells (12 detachable strips of 8), cTnI stock: Lyophilized bovine cTnI (Reconstitute with 0.40mL H20), Enzyme conjugate reagent (11mL), Reference standard (lyophilized): Containing 2μg/mL dog haptoglobin, 20X wash solution (50mL), TMB reagent (11mL), Stop solution (1N HCl): (11mL)
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type HP (Dog) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.