Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Hp (Mouse) ELISA Kit

Catalog No. 89028636 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-636 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-636 Supplier Abnova™ Supplier No. KA1921
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enzyme-linked immunoassay for the quantitative determination of Hp in Mouse Serum or Plasma

Specifications

Calibration Range 1.95 to 125ng/mL
Conjugate HRP
Cross Reactivity Mouse
Description Hp (Mouse) ELISA Kit is a highly sensitive two-site enzyme-linked immunoassay for the quantitative determination of Hp in mouse serum or plasma.
Detection Method Colorimetric
Includes 50mL Diluent concentrate, 50mL wash solution concentrate, 150μL Enzyme Antibody Conjugate concentrate, 12mL TMB Substrate solution, 12mL stop solution, 96-well microtiter plate, 1 vial calibrator (lyophilized)
Quantity 1 Kit
Sample Type Plasma, Serum
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type HP (Mouse) ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.