Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ HPX (Human) ELISA Kit

Catalog No. 89100240 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Milk, Saliva, Urine
Plasma, Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-240 Plasma, Serum
89-100-239 CCS, Milk, Saliva, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-240 Supplier Abnova™ Supplier No. KA0481
Add to Cart
Add to Cart

Includes

  • Human Hemopexin Microplate: A polystyrene microplate coated with a polyclonal antibody against human hemopexin
  • Sealing Tapes: Pressure-sensitive sealing tapes, which can be cut to fit the format of the individual assay
  • Human Hemopexin Standard: Human Hemopexin in a buffered protein base (lyophilized), 20μg
  • Biotinylated Human Hemopexin: Lyophilized
  • MIX Diluent Concentrate (10x)
  • Wash Buffer Concentrate (20x)
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate
  • Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine
  • Stop Solution: A 0.5N hydrochloric acid

This assay employs a quantitative competitive enzyme immunoassay technique that measures human hemopexin in less than 2 hours

Specifications

Calibration Range 0.078 to 5μg/mL
Conjugate Biotin
Cross Reactivity Human
Description HPX (Human) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of human HPX in plasma and serum.
Detection Limit 0.03ug/mL
Detection Method Colorimetric
Includes Antibody coated microwells (96 wells), Human Hemopexin Standard (3.5μg), Biotinylated Hemopexin (1 vial), MIX Diluent Concentrate (10x): 30mL, Wash Buffer Concentrate (20x): 30mL, Streptavidin-Peroxidase Conjugate (SP Conjugate): 80, Chromogen Substrate (8mL), Stop Solution (12mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum
Sample Volume 25 μL
Packaging 96-well
Content And Storage Store the kit at -20°C. The reconstituted standard, SP Conjugate and biotinylated antibody should be stored at -20°C. Others stored at 4°C. Aliquot to avoid repeated freezing and thawing.
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.