Learn More
Description
Includes
- Diluent Concentrate (Running Buffer)
- Wash Solution Concentrate
- Enzyme-Antibody Conjugate 100X
- Chromogen-Substrate Solution: 12mL of 3,3',5,5'-tetramethybenzidine (TMB) and hydrogen peroxide in citric acid buffer at pH 3.3
- Stop Solution: 12mL 0.3M sulfuric acid
- Warning: Avoid contact with skin
- Anti-Rat Hemopexin Elisa Micro Plate: Twelve removable eight (8) well micro well strips in well holder frame. Each well is coated with affinity purified anti-Rat HX
- Rat Hemopexin Calibrator: a lyophilized Rat Hemopexin calibrator
Specifications
Specifications
| Conjugate | HRP |
| Cross Reactivity | Rat |
| Description | Hpx (Rat) ELISA Kit is a two-site enzyme linked immunoassay for the measurement of Hpx. |
| Detection Method | Colorimetric |
| Quantity | 1 Kit |
| Regulatory Status | RUO |
| Sample Type | Biological Fluid |
| Sample Volume | 100 μL |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Quantification |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.