Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ HSP27 (Human) ELISA Kit

Catalog No. 89028038 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-038 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-038 Supplier Abnova™ Supplier No. KA0981
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For detection of Human HSP27 in Plasma, Serum, tissue extract and cell culture lysates

Specifications

Calibration Range 0.078 to 80ng/mL
Conjugate Biotin
Cross Reactivity Human
Description HSP27 (Human) ELISA Kit is designed for detection of human HSP27 in plasma, serum, milk, tissue extract, and cell culture samples.
Detection Limit 0.1ng/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 160ng Human Hsp27 standard, 1 vial Biotinylated Hsp27 Antiody (100X), 30mL EIA diluent concentrate (10X), 30mL x 2 Wash Buffer 20X, 80 Streptavidin-Peroxidase Conjugate (SP Conjugate), 8mL Chromogen Substrate, 12mL stop
Quantity 96 Assays
Sample Type Cell Culture Supernatant, Plasma, Serum, Tissue
Sample Volume 50 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type HSP27 (Human) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.