Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ HSPB1 (Human) ELISA Kit

Catalog No. 89028172 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-172 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-172 Supplier Abnova™ Supplier No. KA1018
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Colorimetric immunometric Enzyme Immunoassay kit for the quantitative measurement of Human Hsp27 in cell lysates, Plasma, Serum and tissue

Specifications

Calibration Range 0.39 to 25ng/mL
Conjugate HRP
Cross Reactivity Human
Description HSPB1 (Human) ELISA Kit is a colorimetric immunometric enzyme immunoassay kit for the quantitative measurement of human Hsp27 in cell lysates, plasma, serum, and tissue.
Detection Limit 0.39ng/mL
Detection Method Colorimetric
Includes 96-well microwell, 5mL EIA Conjugate, 5mL EIA Antibody, 27mL Assay Buffer, 27mL Wash Buffer concentrate, 0.5mL standard, 20mL pNpp Substrate, 5mL stop solution
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Lysate, Plasma, Serum, Tissue
Sample Volume 100 μL
Content And Storage Store control at -20°C. Store all other components at 4°C.
For Use With (Application) Quantification
Product Type HSPB1 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.